Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Thyn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144543.2 | NP_653126.1 | 2, 3 | 196 | ||
GenBank mRNA | AB080370.1 | - | 1, 2 | 120 | ||
AK021335.1 | - | 2, 3 | 164 | |||
AK077639.1 | - | 2, 3 | 166 | |||
BC016594.1 | - | 2, 3 | 150 | 2631627 | ||
CB236103.1 | - | 1, 2 | 389 |