Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam131a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133778.2 | NP_598539.1 | 2, 3 | 268 | ||
GenBank mRNA | AK049983.1 | - | 2, 3 | 269 | ||
BC092532.1 | - | 2, 3 | 299 | 6594874 |