Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wbscr27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024479.2 | NP_077799.2 | 4, 5 | 538 | ||
GenBank mRNA | AK081683.1 | - | 4, 5 | 557 | ||
AK132481.1 | - | 4, 5 | 527 | |||
AY138964.1 | - | 4, 5 | 538 | |||
AY354925.2 | - | 4, 5 | 538 | |||
AY354926.1 | - | 4, 5 | 439 | |||
BC002286.1 | - | 4, 5 | 529 | 3589835 |