Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sp6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031183.2 | NP_112460.1 | 2 | 899 | ||
GenBank mRNA | AJ275988.1 | - | 1 | 128 | ||
AK137686.1 | - | 2 | 1122 | |||
AM295492.1 | - | 2 | 1143 | |||
AY338955.1 | - | 2 | 899 | |||
BC138818.1 | - | 1 | 845 | 8861840 | ||
BC138822.1 | - | 1 | 845 | 8861844 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AK029830.1 | - | 3984 | Chr. - - [-] on Build |