Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Oog3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_201258.2 | NP_957710.2 | 4 | 1450 | ||
GenBank mRNA | AK136030.1 | - | 4 | 1451 | ||
AY573563.1 | - | 4 | 1446 | |||
BC053700.1 | - | 4 | 1446 | 30049894 | ||
BM227552.2 | - | 1 | 400 |