Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BC030499 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001287206.1 | NP_001274135.1 | 12 | 1232 | ||
GenBank mRNA | AK138440.1 | - | 9 | 1641 | ||
BC030499.1 | - | 3 | 480 | |||
BC059191.1 | - | 11 | 1538 |