Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccm2l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145536.3 | NP_663511.2 | 1, 2 | 77 | ||
GenBank mRNA | BC020535.1 | - | 1, 2 | 77 | 4240594 |