Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 2210409E12Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK008869.1 | - | 672 |
Gene Symbol: Gm10367 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK147092.1 | - | 1 | 1394 |
Gene Symbol: Gm15698 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC099439.1 | - | 2 | 347 |