Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rorb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001043354.2 | NP_001036819.1 | 10 | 7415 | ||
NM_001289921.1 | NP_001276850.1 | 11 | 6934 | |||
NM_146095.4 | NP_666207.3 | 10 | 6881 | |||
GenBank mRNA | AK159011.1 | - | 1 | 1683 |