Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Oas1d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133893.3 | NP_598654.1 | 2 | 350 | ||
GenBank mRNA | AB067532.1 | - | 2 | 304 | ||
AK054465.1 | - | 2 | 318 | |||
AK136001.1 | - | 2 | 318 | |||
AY055829.1 | - | 2 | 312 | |||
BC052835.1 | - | 2 | 314 | 30048758 | ||
DV659301.1 | - | 2 | 350 |