Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trrap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081362.1 | NP_001074831.1 | 14 | 1377 | ||
GenBank mRNA | AK147241.1 | - | 14 | 1377 |