Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adamts9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175314.3 | NP_780523.2 | 3 | 314 | ||
GenBank mRNA | CN534419.1 | - | 3 | 314 | ||
CN703716.1 | - | 1 | 18 |