Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: E430018J23Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198011.2 | NP_932128.2 | 3 | 1530 | ||
GenBank mRNA | AK088474.1 | - | 3 | 1494 | ||
BC010442.1 | - | 1528 | 3586722 | |||
BC069858.1 | - | 2 | 1130 |