Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cpne7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_170684.2 | NP_733785.1 | 12 | 1250 | ||
GenBank mRNA | AK032541.1 | - | 3 | 158 | ||
AK049823.1 | - | 9 | 2392 | |||
AK162900.1 | - | 11 | 1869 | |||
BC119328.1 | - | 12 | 1149 | 8734077 | ||
BC137595.1 | - | 12 | 1149 | 8860614 | ||
BM946575.1 | - | 5 | 313 |