Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rpp25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133982.1 | NP_598743.1 | 1 | 375 | ||
GenBank mRNA | AK082822.1 | - | 1 | 445 | ||
AK140177.1 | - | 1 | 451 | |||
BC016085.1 | - | 1 | 375 | 4501535 | ||
BC083064.1 | - | 1 | 435 | 5367203 |