Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sept10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024910.3 | NP_001020081.2 | 5 | 1054 | ||
NM_001024911.2 | NP_001020082.1 | 5 | 1054 | |||
GenBank mRNA | AK076569.1 | - | 5 | 1055 | ||
BC053752.1 | - | 5 | 668 | 6506619 |