Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ttc41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003910.2 | NP_001003910.2 | 2 | 468 | ||
NM_001003939.2 | NP_001003939.2 | 3 | 577 | |||
NM_153595.3 | NP_705823.3 | 1 | 261 | |||
GenBank mRNA | AK133195.1 | - | 2 | 408 | ||
AY651019.1 | - | 4 | 660 | |||
AY651020.1 | - | 1 | 108 | |||
AY651021.1 | - | 1 | 108 | |||
BC030307.1 | - | 3 | 555 |