Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tubg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134024.2 | NP_598785.1 | 11 | 1385 | ||
GenBank mRNA | AB158480.1 | - | 11 | 1397 | ||
AK154019.1 | - | 11 | 1353 | |||
AK160125.1 | - | 11 | 1399 |