Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp692 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040686.1 | NP_001035776.1 | 2, 3 | 375 | ||
NM_182996.3 | NP_892041.2 | 2, 3 | 437 | |||
GenBank mRNA | AK031177.1 | - | 2, 3 | 377 | ||
AK154896.1 | - | 379 | ||||
AK170229.1 | - | 2, 3 | 440 | |||
AK170358.1 | - | 1, 2 | 275 | |||
BC055041.1 | - | 2, 3 | 403 | |||
BC145470.1 | - | 2, 3 | 334 | 9054032 | ||
BC150776.1 | - | 2, 3 | 334 | 9087687 |