Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kdm3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001038695.3 | NP_001033784.2 | 24 | 3754 | ||
NM_173001.3 | NP_766589.1 | 23 | 3781 | |||
GenBank mRNA | AK129204.1 | - | 24 | 3742 | ||
BC005725.1 | - | 3 | 299 | |||
BC026605.1 | - | 9 | 1369 | |||
BC031158.1 | - | 12 | 1841 | |||
BC031200.1 | - | 12 | 1825 | |||
BC031909.1 | - | 7 | 909 | |||
BC049876.1 | - | 17 | 2824 | |||
BC059264.1 | - | 23 | 3664 | 6409698 | ||
DQ323991.1 | - | 20 | 3219 | |||
KJ461739.1 | - | 14 | 2085 |