Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fancm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178912.3 | NP_849243.2 | 14 | 2897 | ||
GenBank mRNA | AK050306.1 | - | 14 | 2898 | ||
AK084697.1 | - | 14 | 2901 | |||
AK173211.1 | - | 14 | 2917 | |||
BC150785.1 | - | 14 | 2891 | 9087696 |