Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tdp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028354.4 | NP_082630.2 | 5, 6 | 814 | ||
GenBank mRNA | AK013427.1 | - | 4, 5 | 289 | ||
AK014855.1 | - | 5, 6 | 794 | |||
AK033868.1 | - | 5, 6 | 812 | |||
BC019804.1 | - | 2, 3 | 138 | 4018534 | ||
BC065162.1 | - | 4, 5 | 784 | 6402921 |