Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Iars | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172015.3 | NP_742012.2 | 3 | 234 | ||
GenBank mRNA | AK087992.1 | - | 3 | 221 | ||
AK133856.1 | - | 3 | 221 | |||
AK136132.1 | - | 3 | 221 | |||
BC067029.1 | - | 3 | 221 | 30534067 | ||
BY139910.1 | - | 3 | 242 |