Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mbnl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175341.4 | NP_780550.1 | 5 | 1333 | ||
NM_207515.2 | NP_997398.1 | 5 | 1333 | |||
GenBank mRNA | AK028797.1 | - | 5 | 1298 | ||
AK085709.1 | - | 5 | 1296 | |||
AK151913.1 | - | 5 | 1297 | |||
AK151926.1 | - | 5 | 1297 | |||
AK151957.1 | - | 5 | 1297 | |||
AK153261.1 | - | 5 | 1187 | |||
AK159213.1 | - | 5 | 1291 | |||
AK164372.1 | - | 5 | 1296 | |||
AK167479.1 | - | 5 | 1163 | |||
AK167694.1 | - | 6 | 1098 | |||
AK220470.1 | - | 1307 | ||||
BC020406.1 | - | 4 | 731 | |||
BC023895.1 | - | 5 | 1285 | |||
BC026711.1 | - | 4 | 698 | |||
BC075665.1 | - | 4 | 936 | 30605146 | ||
BC079898.1 | - | 5 | 1282 | |||
GQ906534.1 | - | 4 | 651 |