Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rcbtb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170694.1 | NP_001164165.1 | 7 | 779 | ||
NM_134083.4 | NP_598844.3 | 7 | 774 | |||
GenBank mRNA | AK031242.1 | - | 7 | 740 | ||
AK031248.1 | - | 7 | 745 | |||
AK147784.1 | - | 8 | 780 | |||
AK160881.1 | - | 8 | 966 | |||
BC003224.1 | - | 5 | 572 | 3482729 | ||
JN086338.1 | - | 6 | 695 | |||
JN086339.1 | - | 7 | 933 |