Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tigd5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178646.4 | NP_848761.2 | 1 | 1282 | ||
GenBank mRNA | AK036241.1 | - | 1 | 1255 | ||
AK051177.1 | - | 1 | 1295 | |||
AK086663.1 | - | 1 | 1259 | |||
AK165093.1 | - | 1 | 1260 | |||
BC099838.1 | - | 1 | 1282 | 5703010 | ||
BC138289.1 | - | 1 | 1256 | 8861308 |