Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lmf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178919.4 | NP_849250.1 | 8 | 1341 | ||
GenBank mRNA | AK042427.1 | - | 8 | 1301 | ||
AK084606.1 | - | 8 | 1167 | |||
AK085435.1 | - | 8 | 1293 | |||
AK149916.1 | - | 8 | 1293 | |||
AK149934.1 | - | 8 | 1342 | |||
AK158371.1 | - | 7 | 1396 | |||
AK171488.1 | - | 8 | 1322 | |||
BC026448.1 | - | 7 | 1121 | |||
BC038039.1 | - | 7 | 1843 | |||
BC058973.1 | - | 6 | 1475 | |||
BC079532.1 | - | 8 | 1237 | 6403184 |