Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mal2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178920.4 | NP_849251.1 | 2, 3 | 433 | ||
GenBank mRNA | AK035986.1 | - | 2, 3 | 315 | ||
BC113154.1 | - | 2, 3 | 310 | 40058489 |