Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Csdc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145473.3 | NP_663448.2 | 3 | 637 | ||
GenBank mRNA | AK028916.1 | - | 3 | 502 | ||
AK081145.1 | - | 1 | 1719 | |||
AK132493.1 | - | 3 | 535 | |||
BC016109.1 | - | 3 | 478 | 4461170 |