Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Csdc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145473.3 | NP_663448.2 | 4 | 791 | ||
GenBank mRNA | AK028916.1 | - | 4 | 656 | ||
AK081145.1 | - | 2 | 1873 | |||
AK132493.1 | - | 4 | 689 | |||
BC016109.1 | - | 4 | 632 | 4461170 |