Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dlk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286013.1 | NP_001272942.1 | 6 | 1114 | ||
NM_001286028.1 | NP_001272957.1 | 6 | 1001 | |||
GenBank mRNA | BC118057.1 | - | 6 | 1001 | 40105170 | |
BC122518.1 | - | 7 | 1029 | 40105175 | ||
BF472059.1 | - | 1 | 110 | |||
BM935328.1 | - | 1 | 19 | |||
FM180474.1 | - | 6 | 1114 |