Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vmac | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166474.1 | NP_001159946.1 | 2 | 326 | ||
NM_178926.3 | NP_849257.1 | 2 | 512 | |||
GenBank mRNA | AK078549.1 | - | 2 | 443 | ||
AK145854.1 | - | 2 | 365 | |||
AK170944.1 | - | 2 | 458 | |||
BC026472.1 | - | 2 | 512 | 4487505 | ||
BC039944.1 | - | 2 | 290 | |||
BC058424.1 | - | 2 | 307 |