Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tnfaip8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177759.1 | NP_001171230.1 | 2 | 501 | ||
NM_001177760.1 | NP_001171231.1 | 2 | 501 | |||
NM_134131.2 | NP_598892.2 | 2 | 639 | |||
GenBank mRNA | AK087478.1 | - | 2 | 558 | ||
AK090316.1 | - | 2 | 449 | |||
AK090350.1 | - | 2 | 525 | |||
AK149901.1 | - | 2 | 415 | |||
AK150445.1 | - | 2 | 411 | |||
AK150965.1 | - | 2 | 421 | |||
AK151622.1 | - | 2 | 411 | |||
AK152426.1 | - | 2 | 411 | |||
AK163339.1 | - | 2 | 400 | |||
BB623947.1 | - | 2 | 501 | |||
BC009090.1 | - | 2 | 401 | 3965693 |