Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lars | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134137.2 | NP_598898.2 | 20 | 2136 | ||
GenBank mRNA | AK129337.1 | - | 20 | 2128 | ||
AK166827.1 | - | 20 | 2124 | |||
BC052715.1 | - | 20 | 2136 | 6836462 |