Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Macrod1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134147.4 | NP_598908.2 | 5 | 734 | ||
GenBank mRNA | AK052998.1 | - | 5 | 693 | ||
BC008653.1 | - | 5 | 459 | 3597023 |