Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Macrod1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134147.4 | NP_598908.2 | 7 | 920 | ||
GenBank mRNA | AI604841.1 | - | 4, 5 | 361 | ||
AK052998.1 | - | 7 | 879 | |||
BC008653.1 | - | 7 | 645 | 3597023 |