Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Carns1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134148.2 | NP_598909.2 | 5 | 588 | ||
GenBank mRNA | AK129347.1 | - | 5 | 563 |