Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vps37c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181403.2 | NP_852068.1 | 5 | 762 | ||
GenBank mRNA | AK030717.1 | - | 4 | 633 | ||
AK150606.1 | - | 5 | 743 | |||
AK158833.1 | - | 4 | 501 | |||
AK159309.1 | - | 5 | 835 | |||
AK165826.1 | - | 5 | 822 | |||
BC025865.1 | - | 5 | 762 | 5029143 |