Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trpt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153597.2 | NP_705825.2 | 5, 6 | 880 | ||
GenBank mRNA | AK038830.1 | - | 4, 5 | 712 | ||
AK048558.1 | - | 1029 | ||||
AK090183.1 | - | 4, 5 | 2011 | |||
AK141582.1 | - | 667 | ||||
BC027575.1 | - | 5, 6 | 678 | 1364225 |