Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gbf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178930.3 | NP_849261.2 | 13 | 1662 | ||
GenBank mRNA | AK039570.1 | - | 13 | 1683 | ||
AK143358.1 | - | 13 | 1674 | |||
AK172919.1 | - | 10 | 1255 | |||
BC004807.1 | - | 8 | 968 | |||
BC011335.1 | - | 13 | 1626 | |||
BC060728.1 | - | 5 | 1082 | |||
BC076569.1 | - | 13 | 1662 | 6409170 | ||
BC082336.1 | - | 14 | 1657 |