Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc25a45 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134154.3 | NP_598915.1 | 3, 4 | 351 | ||
GenBank mRNA | AK133879.1 | - | 3, 4 | 1005 | ||
AK155754.1 | - | 3, 4 | 1004 | |||
AK169876.1 | - | 3, 4 | 997 | |||
AK170869.1 | - | 4, 5 | 482 | |||
BC022156.1 | - | 3, 4 | 182 | |||
BC037680.1 | - | 3, 4 | 355 | 3481846 |