Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc25a45 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134154.3 | NP_598915.1 | 6 | 917 | ||
GenBank mRNA | AK133879.1 | - | 5 | 1690 | ||
AK155754.1 | - | 6 | 1570 | |||
AK169876.1 | - | 6 | 1563 | |||
AK170869.1 | - | 7 | 1048 | |||
BC022156.1 | - | 6 | 748 | |||
BC037680.1 | - | 6 | 921 | 3481846 |