Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acaca | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133360.2 | NP_579938.2 | 43, 44 | 5597 | ||
GenBank mRNA | AF374170.1 | - | 4, 5 | 533 | ||
AY451393.1 | - | 43, 44 | 5571 | |||
BC056500.1 | - | 7, 8 | 1071 |