Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Col16a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028266.5 | NP_082542.3 | 67 | 4508 | ||
GenBank mRNA | AK031163.1 | - | 58 | 3373 | ||
AK040971.1 | - | 67 | 4461 | |||
BC023940.1 | - | 21 | 1217 | |||
BC027766.1 | - | 67 | 4443 | 5346676 |