Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Krt9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_201255.2 | NP_957707.2 | 4, 5 | 1011 | ||
GenBank mRNA | AY497550.1 | - | 4, 5 | 1011 |