Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pacs1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153129.2 | NP_694769.2 | 10, 11 | 1522 | ||
GenBank mRNA | AK154563.1 | - | 1395 | |||
AK164770.1 | - | 11, 12 | 1508 | |||
AK171969.1 | - | 10, 11 | 1505 | |||
BC062886.1 | - | 10, 11 | 1522 | 5706409 | ||
CG671741.1 | - | 2, 3 | 244 |