Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ap4e1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175550.3 | NP_780759.2 | 18, 19 | 2912 | ||
GenBank mRNA | AK036944.1 | - | 18, 19 | 2893 | ||
BC042530.1 | - | 5, 6 | 1103 |