Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ap4e1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175550.3 | NP_780759.2 | 5 | 507 | ||
GenBank mRNA | AK036944.1 | - | 5 | 489 | ||
AK039257.1 | - | 5 | 495 | |||
AK143351.1 | - | 5 | 497 | |||
AK144636.1 | - | 5 | 487 |