Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnf144a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081977.2 | NP_001075446.1 | 7 | 856 | ||
NM_080563.4 | NP_542130.1 | 6 | 782 | |||
GenBank mRNA | AF360998.1 | - | 7 | 852 | ||
AK038586.1 | - | 6 | 674 | |||
AK133544.1 | - | 4 | 396 | |||
AK172904.1 | - | 5 | 617 | |||
BC030187.1 | - | 6 | 746 | 5345434 | ||
BQ884505.1 | - | 6 | 715 |