Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Galnt16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081421.1 | NP_001074890.1 | 13 | 1495 | ||
GenBank mRNA | AB045325.1 | - | 13 | 1457 | ||
AB175683.1 | - | 13 | 1316 | |||
AK138883.1 | - | 13 | 1450 | |||
AK173105.1 | - | 14 | 1607 | |||
BC110634.1 | - | 14 | 1590 | 40059147 | ||
BC125015.1 | - | 14 | 1589 | 40059141 |
Gene Symbol: 4933406B15Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK016679.1 | - | 1 | 12 |